missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone, 98%

Catalog No. p-3208360
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1759003 1 g
AAA1759006 5 g
AAA1759014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1759003 Supplier Thermo Scientific Chemicals Supplier No. A1759003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-02-2 | C22H29FO5 | 392.47 g/mol

Anti-inflammatory glucocorticoidDexamethasone is used to treat inflammatory and autoimmune conditions such as rheumatoid arthritis and bronchospasm. It is useful to study apoptosis, cell signaling pathways and gene expression. It is associated with marbofloxacin and clotrimazole and finds application in veterinary medicine to treat difficult ear infections in dogs. It is also used to treat horses with swelling of of distal limbs and general bruising in combination with trichlormethiazide. It is also an anti-inflammatory glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anti-inflammatory glucocorticoidDexamethasone is used to treat inflammatory and autoimmune conditions such as rheumatoid arthritis and bronchospasm. It is useful to study apoptosis, cell signaling pathways and gene expression. It is associated with marbofloxacin and clotrimazole and finds application in veterinary medicine to treat difficult ear infections in dogs. It is also used to treat horses with swelling of of distal limbs and general bruising in combination with trichlormethiazide. It is also an anti-inflammatory glucocorticoid.

Solubility
Soluble in ethanol and dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.47
MDL Number MFCD00064136
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
Synonym dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
PubChem CID 5743
ChEBI CHEBI:41879
IUPAC Name (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Melting Point ∽260°C (decomposition)
Assay Percent Range 0.98
Quantity 1 g
Beilstein 2066651
Sensitivity Light sensitive
Merck Index 142943
Solubility Information Soluble in ethanol and dimethyl sulfoxide. Insoluble in water.
Formula Weight 392.47
Percent Purity 98%
Chemical Name or Material Dexamethasone
Hazard Category Carcinogenicity (category 2)
Hazard Statement Suspected of causing cancer
EINECSNumber 200-003-9
RTECSNumber TU3980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only