missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DeviceLink Connect Hardware

Catalog No. DLCPTBRKT1
Change view
Click to view available options
Description:
Cable mounting hardware (spare parts)
DeviceLink Connect mote (only)
Ethernet (LAN) cable
LN2 Temperature sensor mounting bracket
USB Commissioning (setup) dongle
For Use With (Equipment):
DeviceLink Connect mote
DeviceLink Connect system
LN2 Cryo chambers
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment)
DLCPTBRKT1 LN2 Temperature sensor mounting bracket LN2 Cryo chambers
DLCPTETHE3 Ethernet (LAN) cable DeviceLink Connect mote
DLCPTMONT1 Cable mounting hardware (spare parts) DeviceLink Connect system
DLCPTMOTE1 DeviceLink Connect mote (only) DeviceLink Connect system
DLCSWCOMM1 USB Commissioning (setup) dongle DeviceLink Connect mote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. DLCPTBRKT1 Supplier Thermo Scientific™ Supplier No. DLCPTBRKT001

The DeviceLink Connect is a remote monitoring and asset management solution for laboratory equipment enabling knowledge based decisions by monitoring critical equipment parameters while providing data insights and real-time system notifications.

Thermo Scientific™ DeviceLink Connect is the newest addition to Thermo Scientific’s DeviceLink portfolio that measures critical reliability parameters and uses advanced analytics to provide real-time monitoring and reporting for lab instruments. The system supports independent sensors to monitor your instrument, but it can also be connected via serial port directly to the instrument harnessing its on-board telemetry data, alarms, and events. All information is transmitted to the Thermo Connect cloud where you have 24/7 access to the information via web and mobile based platforms.

The DeviceLink Connect is a remote monitoring system that uses direct serial/USB based instrument connections and/or independent sensors to gather useful information about the operation of the equipment. Sending this data back to Thermo Connect cloud allows for 24/7 monitoring from anywhere in the world.

How it works:

  1. Connect your DeviceLink Connect system
    • Supply power via the provided AC power cable
    • Connect your instrument via serial/USB cable to the DeviceLink Connect mote
    • Install (optional) independent sensors
  2. Configure your setup via the web-based Commissioner Application
  3. Establish a Thermo Connect cloud account
  4. Download the free mobile InstrumentConnect mobile application
  5. Monitor your equipment 24/7 from anywhere in the world

Network:

  • Wi-Fi: WPA Enterprise (802.1x) and WPA2-PSK protocols
  • Ethernet (LAN) connections also supported
  • Up to 7 days internal data storage during a network outage

Power and Battery:

  • Power-over-Ethernet (PoE) provides main power to the device.
  • PoE wall adapter provide or a PoE 802.3af standard connection can be used
  • Contains a rechargeable Li-ion backup battery providing up to 3 hrs. of backup power during continuous operation (2600 mAH, 3.6V)

Advanced InSight Analytics:

The DeviceLink Connect system is also compatible with Thermo Scientific™ InSight™ analytics, powered by KLATU™ which are designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:

  • Cabinet temperature
  • Electrical use and energy consumption
  • Door openings
  • Ambient temperature

When you purchase an InSight monitoring license and subscription (sold separately) your DeviceLink Connect data run through a number of algorithms utilizing the InSight software. With InSight analytics, you can determine and compare the assets' performance against industry performance standards adding that next level of asset management.

Installation and Services:

  • Customer (self) installation
  • Install services are sold separately if desired
  • Calibration sensors also available.

Available only in North America.

Specifications

Description LN2 Temperature sensor mounting bracket
For Use With (Application) Wireless Monitoring, Asset Management Solution
For Use With (Equipment) LN2 Cryo chambers