missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Development Folders (14 cm X 19 cm)

Catalog No. T2258
Encompass
Change view
Click to view available options
Quantity:
30 folders
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2258 30 folders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2258 Supplier Applied Biosystems™ Supplier No. T2258

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use during imaging of blotting membranes following chemiluminescent signal development (western blotting, nucleic acid blotting).

  • Size: 14 x 19cm

Specifications

Content And Storage Store at room temperature.
Quantity 30 folders
Shipping Condition Room Temperature
Product Type Development Folder

For Research Use Only. Not for use in diagnostic procedures.