Learn More
Research Products Corporation Detojet Low Foaming Liquid Detergent, 1 Gallon
Catalog No. 501367628
Encompass
Detojet is a low foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems, and ultrasonic cleaning systems. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no interfering residues. Protects interior working parts of washers. USDA authorized. Dilute 1:200. pH 12.4
Concentrated to save you money
Biodegradable and disposable
Low foaming heavy duty detergent for high performance results from your cleaning machinery
Free rinsing to give you reliable results and no interfering residues
Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
Catalog No. 501367628 Supplier Research Products Corporation Supplier No. 114029
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Detojet is a low foaming, heavy duty alkaline detergent for use in labware washers, parts washers, power-spray systems, and ultrasonic cleaning systems. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no interfering residues. Protects interior working parts of washers. USDA authorized. Dilute 1:200. pH 12.4
Concentrated to save you money
Biodegradable and disposable
Low foaming heavy duty detergent for high performance results from your cleaning machinery
Free rinsing to give you reliable results and no interfering residues
Use to pass your cleaning validation tests for lab accreditation and plant inspection approval