missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Detachable Cables

Accessories for use with electrodes.

$460.36

Specifications

Color Gray
Connector Types SC-LEMO
For Use With (Equipment) Amperometric Titration and Double Pin Platinum Electrode
Type Cable
View More Specs

Products 1
Catalog Number Mfr. No. Length (Metric) Cable Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Cable Price Quantity & Availability  
01910742
View Documents Promotion Details
Mettler Toledo™
51109184
1 m
Each for $460.36
Only null left
 
Description

Description

For use with electrodes.
Specifications

Specifications

Gray
SC-LEMO
Amperometric Titration and Double Pin Platinum Electrode
Cable
Videos
SDS
Documents

Documents