missing translation for 'onlineSavingsMsg'
Learn More

Carson™ DeskBrite 300 2x COB LED Lighted 3.5 in. Desktop Magnifier

Catalog No. S81722
Encompass
Change view
Click to view available options
Magnification:
2x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S81722 2x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81722 Supplier Carson™ Supplier No. LM30

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DeskBrite 300 includes 5x spot lens and USB-C wall adapter, adjustable flexible neck allows users to position lens

  • Contains an aspheric design
    • Light rays pass through lens and meet at a single point allowing for crystal clear, distortion-free image quality
  • Magnifier power by USB-C to wall adapter (included) or 3 AA batteries (not included)
  • For reading fine print, hobbies, crafts, sewing, needlework, model building, jewelry making, soldering, painting nails, inspection
  • Back by Carson™'s limited lifetime warranty

Includes:

  • Four ultra bright COB LED lights with two brightness settings

Specifications

Product Type Magnifier, Desktop
Material Acrylic
Diameter (Englis) Lens 3.5 in.
Diameter (Metric) Lens 0.089 m
Magnification 2x