Learn More
DWK Life Sciences KIMBLE™ Desiccator with Plastic Top Knob, 150x255
Catalog No. 12141430
Change view
Click to view available options
Product Type:
Desiccator with Plastic Top Knob
Catalog No. Product Type
12141430 Desiccator with Plastic Top Knob
1 options
Catalog No. 12141430 Supplier DWK Life Sciences Supplier No. 31050150
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The KIMBLE™ Desiccator with Plastic Top Knob, 150x255 is ideal for storage of hygroscopic chemicals and prevents moisture adsorption when using crucibles, weighing bottles, and watch glasses. Features a plastic screw-thread top cap and recessed base for desiccant containment.
  • Plastic screw-thread top cap for easy cover removal
  • Recessed base for containment of desiccant
  • Includes metal plate
  • Manufactured from soda-lime glass conforming to USP Type III and ASTM E438, Type II, requirements
Specifications
Material Glass
Product Type Desiccator with Plastic Top Knob
Height (Metric) 255 mm
Diameter (Metric) Inner 150 mm