Learn More
Worthington Biochemical DEOXYRIBONUCLEASE I RN 100UN
Catalog No. 50592516
Encompass
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Supplied as a solution at approximately 2 Kunitz units per microliter (approximately 1 mg/ml) containing 50% glycerol and 1 mM calcium chloride. 9003-98-9
Catalog No. 50592516 Supplier Worthington Biochemical Supplier No. LS006342
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Molecular Biology Grade. Chromatographically purified to remove RNase and protease. Supplied as a solution at approximately 2 Kunitz units per microliter (approximately 1 mg/ml) containing 50% glycerol and 1 mM calcium chloride. 9003-98-9