Learn More
Worthington Biochemical DEOXYRIBONUCLEASE I, RECOMBINA
Catalog No. NC0527673
Encompass
Recombinant Bovine pancreatic Deoxyribonuclease 1 produced in Pichia pastoris. Chromatographically purified. Free of animal derived components, RNases, and proteases. A lyophilized powder containing glycine as a stabilizer.
Catalog No. NC0527673 Supplier Worthington Biochemical Supplier No. LS006361
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Bovine pancreatic Deoxyribonuclease 1 produced in Pichia pastoris. Chromatographically purified. Free of animal derived components, RNases, and proteases. A lyophilized powder containing glycine as a stabilizer.