missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Deoxycholic Acid, Sodium Salt, ≥96%, Calbiochem™,
Chemical Identifiers
| CAS | 302-95-4 |
|---|---|
| Molecular Formula | C24H39NaO4 |
| Molecular Weight (g/mol) | 414.56 |
| MDL Number | MFCD00064139 |
| InChI Key | FHHPUSMSKHSNKW-SMOYURAASA-M |
| PubChem CID | 91810855 |
| IUPAC Name | sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| SMILES | [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
Chemical Identifiers
| 302-95-4 | |
| 414.56 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| C24H39NaO4 | |
| MFCD00064139 | |
| 91810855 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C |
Specifications
| White | |
| C24H39NaO4 | |
| FHHPUSMSKHSNKW-SMOYURAASA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate | |
| 91810855 |
| 302-95-4 | |
| MFCD00064139 | |
| [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C | |
| 414.56 | |
| ≥96% (TLC) |