missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic acid, 99%, Thermo Scientific Chemicals

Catalog No. aab2006114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2006114 25 g
AAB2006122 100 g
AAB2006136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2006114 Supplier Thermo Scientific Chemicals Supplier No. B2006114
Only null left

Deoxycholic acid has been used in a modified procedure to recover 40-80% of a protein from a 1 μg/mL solution. It forms complexes with fatty acid. Used as an emulsifying agent in food, a precursor in the synthesis of cortisone, and a gallbladder stimulant. It has been used to study assess how physiological concentrations of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA) affect barrier function in mouse intestinal tissue. Deoxycholic acid has been used in a study to assess a pH-Responsive Mechanism of a Deoxycholic Acid and Folate Co-Modified Chitosan Micelle under Cancerous Environment. It has also been used in a study to investigate dose-dependent anti-inflammatory effect of ursodeoxycholic acid in experimental colitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxycholic acid has been used in a modified procedure to recover 40-80% of a protein from a 1 µg/mL solution. It forms complexes with fatty acid. Used as an emulsifying agent in food, a precursor in the synthesis of cortisone, and a gallbladder stimulant. It has been used to study assess how physiological concentrations of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA) affect barrier function in mouse intestinal tissue. Deoxycholic acid has been used in a study to assess a pH-Responsive Mechanism of a Deoxycholic Acid and Folate Co-Modified Chitosan Micelle under Cancerous Environment. It has also been used in a study to investigate dose-dependent anti-inflammatory effect of ursodeoxycholic acid in experimental colitis.

Solubility
The free acid is soluble at 0.044 g/L in water at 20°C. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 83-44-3
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003673
InChI Key KXGVEGMKQFWNSR-LLQZFEROSA-N
Synonym deoxycholic acid, deoxycholate, desoxycholic acid, choleic acid, cholerebic, cholorebic, degalol, deoxycholatic acid, droxolan, pyrochol
PubChem CID 222528
ChEBI CHEBI:28834
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 172°C to 176°C
Quantity 25 g
Beilstein 3219882
Merck Index 14,2899
Solubility Information The free acid is soluble at 0.044g/L in water at 20°C. Solubility in methanol is almost transparent.
Optical Rotation +54° (c=2 in Ethanol)
Formula Weight 392.58
Percent Purity 99%
Chemical Name or Material Deoxycholic acid
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-478-5
RTECSNumber FZ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only