missing translation for 'onlineSavingsMsg'
Learn More

Deoxybenzoin, 97%

Catalog No. AC111960250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111960250 25 g
AC111961000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111960250 Supplier Thermo Scientific Chemicals Supplier No. 111960250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 451-40-1 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 451-40-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00003081
InChI Key OTKCEEWUXHVZQI-UHFFFAOYSA-N
Synonym 2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
PubChem CID 9948
IUPAC Name 1,2-diphenylethanone
SMILES O=C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52.0°C to 57.0°C
Color White
Boiling Point 320.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CH2COC6H5
Quantity 25 g
Beilstein 07, II, 368
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohols and ketones
Formula Weight 196.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Deoxybenzoin, 98%
Show More Show Less
EINECSNumber 207-193-2
RTECSNumber AM9662500
TSCA TSCA

RUO – Research Use Only