missing translation for 'onlineSavingsMsg'
Learn More

Deoxybenzoin, 97%

Catalog No. AAA1058022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1058022 100 g
AAA1058014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1058022 Supplier Thermo Scientific Chemicals Supplier No. A1058022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 451-40-1 | C14H12O | 196.25 g/mol

Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxybenzoin, is used as a benzoin derivative and as a photoinitiator in vinyl polymerization.

Solubility
Soluble in water (partly), methanol, alcohols, and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions.

Chemical Identifiers

CAS 451-40-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00003081
InChI Key OTKCEEWUXHVZQI-UHFFFAOYSA-N
Synonym 2-phenylacetophenone, deoxybenzoin, benzyl phenyl ketone, ethanone, 1,2-diphenyl, desoxybenzoin, 1,2-diphenylethan-1-one, phenyl benzyl ketone, benzoin, deoxy, acetophenone, 2-phenyl, deoxy benzoin
PubChem CID 9948
IUPAC Name 1,2-diphenylethanone
SMILES O=C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 53°C to 57°C
Density 1.2
Boiling Point 319°C to 321°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1072876
Solubility Information Soluble in water (partly),methanol,alcohols,and ketones.
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Deoxybenzoin
EINECSNumber 207-193-2
RTECSNumber AM9662500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only