Learn More
Eisco™ Density Spheres
The Eisco Labs Copper Hollow Sphere has a 75 mm diameter. It demonstrates the effect of temperature on liquid density.
Supplier:  Eisco™ PH0113C
Numéro de catalogue. S02913
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Copper hollow sphere
  • Demonstrates the effect of temperature on the density of a liquid
  • Diameter 75 mm
  • Weighted - will float incool water 15°C and sink in water 30°C
Spécifications
Spécifications
Density Ball
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS