missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Density Rod

Catalog No. S15844
Change view
Click to view available options
Product Type:
Density Demonstration Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S15844 Density Demonstration Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15844 Supplier United Scientific™ Supplier No. DNROD1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Classic demonstration that engages students.

  • Machined aluminum cylinder floats in cold water but sinks in warm water.
  • Ideal for classroom demonstrations around density
  • Includes activity guide

Specifications

For Use With (Application) Beaker
Product Type Density Demonstration Kit

Warning: Cancer and reproductive harm