missing translation for 'onlineSavingsMsg'
Learn More

DenLine Protection Plus Unisex 41 in. (104 cm) Long Open Back Gown- Fluid Resistant Barrier

Catalog No. 22412099
Encompass
Change view
Click to view available options
Color:
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
22412099 White 3X-Large
22412089 White Small
22412093 White Large
22412091 White Medium
22412095 White X-Large
22412097 White 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22412099 Supplier DenLine Protection Plus Supplier No. DL1753XLWHT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Open Back Gown with One Piece Seamless Front and High Neck Coverage provides Wearer's Front, Shoulders Level 2 Protection from Incidental splash of Blood, Body Fluids, Aerosols, etc.; Open Back provides Exceptional Comfort to Wearer; Stain Resistant, Anti-static; 225 Commercial Wash Tested.

Primary Applications

· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics

Wearer Protection Provided

  • One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
  • Adjustable snaps at neck for complete coverage. Back ties at waist.
  • No patch pockets on front for splash of blood, body fluids, aerosols or OPIM to accumulate.
  • Open Back Design offers protection to shoulders from splash traveling either side of employee’s face.
  • AAMI PB70- Level 2, Assists wearers comply with OSHA 29 CFR 1910

Wearer Comfort

  • Overall Air Permeability of Front and Arms 12 CFM (ASTM D737)…Open Back
  • Raglan Sleeves and Loose Fit contribute to wearer comfort.

Other Features

  • Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
  • Anti-static Fabric prevents false instrument readings, damage to computers
  • Lint Free Fabric prevents sample contamination, ideal for molecular testing
  • Durable Construction
    • Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
    • Durable Cuffs for Overgloving – Bleach Safe all colors
  • Choice of 2 Colors, Ceil Blue and White; Sizes S to 3XL
  • 225 Commercial Wash Tested; Low Lifetime Cost per Wash
  • Autoclavable Fabric (274°F/135°C).
  • Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen

Specifications

Size 3X-Large
Material 99% Textured Polyester, 1% Carbon
Color White
Disposable Reusable
Closure Type Snap, Ties
Cuff Style Knit
Collar Type High
Finish Fluid and Stain Resistant, Anti-Static
Size (English) Chest 54 to 56 in.
Size (Metric) Chest 137 to 142 cm
Certifications/Compliance OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
Length (Metric) 104.1 cm
Length (English) Cuff 4 in.
Length (Metric) Cuff 10.1 cm
Weight (English) 0.6 lb.
Autoclavable Autoclavable
Permeability Overall Air Permeability 12 CFM (ASTM D737)
Construction Fluid Resistant Front and Shoulders
Length (English) 41 in.
For Use With (Application) Protection Against Incidental Splash of Blood & OPIM
Show More Show Less