Learn More
DenLine Protection Plus Unisex 41 in. (104 cm) Long Open Back Gown- Fluid Resistant Barrier
Open Back Gown with One Piece Seamless Front and High Neck Coverage provides Wearer's Front, Shoulders Level 2 Protection from Incidental splash of Blood, Body Fluids, Aerosols, etc.; Open Back provides Exceptional Comfort to Wearer; Stain Resistant, Anti-static; 225 Commercial Wash Tested.
Supplier: DenLine Protection Plus DL1752XLWHT
Description
Primary Applications
· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics
Wearer Protection Provided
- One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
- Adjustable snaps at neck for complete coverage. Back ties at waist.
- No patch pockets on front for splash of blood, body fluids, aerosols or OPIM to accumulate.
- Open Back Design offers protection to shoulders from splash traveling either side of employee’s face.
- AAMI PB70- Level 2, Assists wearers comply with OSHA 29 CFR 1910
Wearer Comfort
- Overall Air Permeability of Front and Arms 12 CFM (ASTM D737)…Open Back
- Raglan Sleeves and Loose Fit contribute to wearer comfort.
Other Features
- Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
- Anti-static Fabric prevents false instrument readings, damage to computers
- Lint Free Fabric prevents sample contamination, ideal for molecular testing
- Durable Construction
- Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
- Durable Cuffs for Overgloving – Bleach Safe all colors
- Choice of 2 Colors, Ceil Blue and White; Sizes S to 3XL
- 225 Commercial Wash Tested; Low Lifetime Cost per Wash
- Autoclavable Fabric (274°F/135°C).
- Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen
Specifications
| 2X-Large | |
| White | |
| Snap, Ties | |
| Protection Against Incidental Splash of Blood & OPIM | |
| Fluid and Stain Resistant, Anti-Static | |
| 127 to 132 cm | |
| 41 in. | |
| 4 in. | |
| 0.6 lb. | |
| Overall Air Permeability 12 CFM (ASTM D737) |
| 99% Textured Polyester, 1% Carbon | |
| Reusable | |
| Knit | |
| High | |
| 50 to 52 in. | |
| OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection | |
| 104.1 cm | |
| 10.1 cm | |
| Autoclavable | |
| Fluid Resistant Front and Shoulders |