missing translation for 'onlineSavingsMsg'
Learn More

DenLine Protection Plus Unisex 41 in. (104 cm) Long Open Back Gown- Fluid Resistant Barrier

Open Back Gown with One Piece Seamless Front and High Neck Coverage provides Wearer's Front, Shoulders Level 2 Protection from Incidental splash of Blood, Body Fluids, Aerosols, etc.; Open Back provides Exceptional Comfort to Wearer; Stain Resistant, Anti-static; 225 Commercial Wash Tested.

Supplier:  DenLine Protection Plus DL175XLWHT

 View more versions of this product

Catalog No. 22-412-095

Only null left
Explore available promotions

Description

Description

Primary Applications

· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics

Wearer Protection Provided

  • One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
  • Adjustable snaps at neck for complete coverage. Back ties at waist.
  • No patch pockets on front for splash of blood, body fluids, aerosols or OPIM to accumulate.
  • Open Back Design offers protection to shoulders from splash traveling either side of employee’s face.
  • AAMI PB70- Level 2, Assists wearers comply with OSHA 29 CFR 1910

Wearer Comfort

  • Overall Air Permeability of Front and Arms 12 CFM (ASTM D737)…Open Back
  • Raglan Sleeves and Loose Fit contribute to wearer comfort.

Other Features

  • Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
  • Anti-static Fabric prevents false instrument readings, damage to computers
  • Lint Free Fabric prevents sample contamination, ideal for molecular testing
  • Durable Construction
    • Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
    • Durable Cuffs for Overgloving – Bleach Safe all colors
  • Choice of 2 Colors, Ceil Blue and White; Sizes S to 3XL
  • 225 Commercial Wash Tested; Low Lifetime Cost per Wash
  • Autoclavable Fabric (274°F/135°C).
  • Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen
Specifications

Specifications

X-Large
White
Snap, Ties
Protection Against Incidental Splash of Blood & OPIM
Fluid and Stain Resistant, Anti-Static
117 to 122 cm
41 in.
4 in.
0.6 lb.
Overall Air Permeability 12 CFM (ASTM D737)
99% Textured Polyester, 1% Carbon
Reusable
Knit
High
46 to 48 in.
OSHA 29 CFR 1910, AAMI PB70 Level 2 Protection
104.1 cm
10.1 cm
Autoclavable
Fluid Resistant Front and Shoulders
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions