missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DenLine Protection Plus Unisex 41 in. (104 cm) Long Closed Back Gown- Fluid Resistant Barrier (AAMI Level 2)
Closed back gown provides AAMI Level 2 protection from incidental splash of blook, bodily fluids, and aerosols.
Supplier: DenLine Protection Plus DL176LMWHT
Description
Primary Applications
· Anatomic Pathology · Clinical Pathology · Clinical Laboratory · Phlebotomy · Specimen Collection · Blood Collection, Testing, & Manufacturing · Human & Animal Research · Academia · Dialysis · Dental · Clean Room ISO 5/Class 100 - Medical Device, Pharma, Electronics
Wearer Protection Provided
- One Piece Seamless front protects wearers from neck to below knees, arms, and covers tops of legs with no gaping. Textured Fabric holds fabric into position on top of legs when seated.
- Adjustable Snaps at Neck for full Coverage. Back Ties at Waist
- No patch pockets on front for splash of blood or OPIM to accumulate.
- Closed Back Design offers protection to shoulders and back from splash traveling either side of employee’s face.
- AAMI PB7- Level 2, Assists wearers comply with OSHA 29 CFR 1910:
Wearer Comfort
- Overall Air Permeability 12 CFM (ASTM D737)
- Raglan Sleeves and Loose Fit contribute to wearer comfort.
Other Features
- Hidden Side Seam Accessible Inside Pockets (2), protected from frontal splash.
- Stain Resistant Fabric and Cuffs (blood, bleach and all aqueous reagents)
- Anti-static Fabric prevents false instrument readings, damage to computers
- Low-Lint Fabric prevents sample contamination, ideal for molecular testing
- Badge Tag Loop
- Durable Construction
- Stainless Snap Closures at Back of Neck – Lifetime Guaranteed
- Durable Cuffs for Overgloving – Bleach Safe all colors
- Choice of 2 Colors, Royal and White, Sizes XS to 3XL
- 225 Commercial Wash Tested; Low Lifetime Cost per Wash
- Autoclavable Fabric (274°F/135°C).
- Launder using Home/Laundromat or Commercial Equipment; Instructions furnished meet CDC/HICPAC Recommendations for processing medical bloodborne Linen
Specifications
| 41 in. | |
| General Purpose Gown | |
| None | |
| Closed Back Gown | |
| Medium | |
| White | |
| Reusable | |
| Knit | |
| Yes | |
| 0.6 lb. | |
| Overall Air Permeability 12 CFM (ASTM D737) |
| 360° Fluid Resistance Front, Shoulders, and Back | |
| Protection Against Incidental Splash of Blood & OPIM | |
| Unisex | |
| 104.1 cm | |
| Textured Polyester (99%), Carbon (1%) | |
| Adjustable Snap at Neck, Back Ties at Waist | |
| Non-sterile | |
| Fluid and Stain Resistant, Anti-Static | |
| OSHA 29 CFR 1910, AAMI PB70 Level 2 | |
| Autoclavable |