missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Demonstration Electroscope

Catalog No. S27748
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27748 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27748 Supplier Eisco™ Supplier No. PH0917A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Demonstration Electroscope designed to demonstrate static potentials. Includes a charging ball and set of condenser plates with an insulated rod.

  • Ring Diameter: 150 mm

Specifications

Grade 7 to Undergraduate
For Use With (Application) Demonstrating Static Potentials
Includes Charging Ball, and a Condenser Plate Set with an Insulated Rod
Quantity 1 Ea.
Class Size 1 Class
Product Type Demonstration Electroscope