Learn More
Brady™ Deluxe Visitor Spec Dispenser
Catalog No. p-7184625
Change view
Click to view available options
Color:
Clear
Fluorescent Green
Fluorescent Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19129474 Clear
19129475 Fluorescent Green
19129478 Fluorescent Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19129474 Supplier Brady™ Supplier No. MVSD
Only null left
Eyewear dispensers remind employees to wear protective equipment and save much-needed room by storing the equipment in space-efficient containers.
  • With bottom door that keeps glasses in and folds down for easy dispensing
  • Each dispenser holds approximately 20 to 25 glasses
  • Dispensers are supplied with two top holes making them ideal for wall mounting
Specifications
Color Clear
Product Type Safety Glasses Dispenser (Unfilled)
Size (H x W x D) 18 x 8 x 4 in.
Material Acrylic