missing translation for 'onlineSavingsMsg'
Learn More

Deluxe Van de Graaff Generator

Demonstrates electrostatic phenomena.

$61.64 - $2,785.82

Specifications

For Use With (Application) Demonstrating static electricity
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S43207
Accessories View Documents Promotion Details
N/A Van de Graaff Generator
Each for $2,785.82
Only null left
 
S43208
View Documents Promotion Details
N/A Replacement Belt
Each for $61.64
Only null left
 
Description

Description

A first-rate apparatus for demonstrating static electricity. 

  • Generates potentials as high as 400,000 volts
  • Produces sparks as long as 15 in. (38cm)
  • Belt current charges the dome at 10μA; replacement belts are available
  • Operates on 110VAC with grounded cord
  • Rotary Dial located in the unit's base allows you to vary the motor's speed
  • Terminal located on the 9.8 × 11.8 in. (25 × 30cm) base is grounded to safely produce sparks and discharge the dome
  • Aluminum Sphere is 9.8 in. (25cm) diameter; creates sparks of great intensity
  • Aluminum electrode (sold separately) is supported by an insulated, polycarbonate column
  • Overall height: 29.9 in. (76cm)
Specifications

Specifications

Demonstrating static electricity
Videos
SDS
Documents

Documents