Learn More
United Scientific™ Deluxe Boyle's Law Apparatus
Catalog No. S012432
Change view
Click to view available options
Dimensions (L x W x H):
6 x 4.5 x 10 in.
Catalog No. Dimensions (L x W x H)
S012432 6 x 4.5 x 10 in.
1 options
Cancer and reproductive harm.
Catalog No. S012432 Supplier United Scientific™ Supplier No. BYLAPVT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cancer and reproductive harm.
Explore the relationship between pressure, volume, and temperature with this Boyle’s Law Apparatus, featuring a built-in thermocouple to monitor air temperature and a tube connected to an absolute pressure gauge.
  • Cylinder has mL markings, and the screw mechanism allows you to adjust the volume and keep it at a specific position
  • Can close the air vent and adjust the volume of the cylinder, allowing you to measure the changes in pressure and temperature as you increase or decease the volume
  • Built in digital thermometer
  • Compact design
  • Easy-to-read pressure gauge
  • One AA battery required
Specifications
For Use With (Application) Determination of Gas Pressure and Volume In Closed Container
Includes Built-in Digital Thermometer, Cylinder, Pressure Gauge
Dimensions (L x W x H) 6 x 4.5 x 10 in.
Product Type Boyle's Law Demonstration Apparatus