missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactam, 98%

Catalog No. AAA1227122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1227122 100 g
AAA1227114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1227122 Supplier Thermo Scientific Chemicals Supplier No. A1227122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 675-20-7 | C5H9NO | 99.13 g/mol

delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 675-20-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006037
InChI Key XUWHAWMETYGRKB-UHFFFAOYSA-N
Synonym 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
PubChem CID 12665
ChEBI CHEBI:77761
IUPAC Name piperidin-2-one
SMILES O=C1CCCCN1

Specifications

Melting Point 35°C to 39°C
Boiling Point 255°C to 257°C
Flash Point 147°C (296°F)
Quantity 100 g
Beilstein 106434
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 99.13
Percent Purity ≥98%
Chemical Name or Material delta-Valerolactam
Hazard Category H319-H335
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 211-622-9
RTECSNumber TO0110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only