Learn More
Waters Limited DeltaPak C18 Radial-Pak Guard, 300, 15 m, 25 mm X 10 mm, 2/pk
Catalog No. 50785466
Encompass
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5m or 15m spherical silica. Delta-Pak is available in two different pore sizes (100 and 300) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5m particle, you can easily check the purity of your fractions.
Catalog No. 50785466 Supplier Waters Limited Supplier No. WAT038522
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5m or 15m spherical silica. Delta-Pak is available in two different pore sizes (100 and 300) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5m particle, you can easily check the purity of your fractions.