missing translation for 'onlineSavingsMsg'
Learn More

delta-Nonanolactone 98.0+%, TCI America™

Catalog No. N042425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N042425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N042425ML Supplier TCI America Supplier No. N042425ML
Only null left

Chemical Identifiers

CAS 3301-94-8
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036500
InChI Key PXRBWNLUQYZAAX-UHFFFAOYSA-N
Synonym delta-nonalactone, 5-nonanolide, 6-butyltetrahydro-2h-pyran-2-one, 2h-pyran-2-one, 6-butyltetrahydro, 5-nonalactone, delta-nonanolactone, hydroxynonanoic acid delta-lactone, 5-butyl-delta-valerolactone, delta-butyl-delta-valerolactone, delta-hydroxypelargonic acid lactone
PubChem CID 18698
IUPAC Name 6-butyloxan-2-one
SMILES CCCCC1CCCC(=O)O1

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 137°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Nonanolactone
EINECSNumber (5)-0698
TSCA Yes