missing translation for 'onlineSavingsMsg'
Learn More

delta-Hexanolactone, 98%

Catalog No. AAA1927422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1927422 100 g
AAA1927414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1927422 Supplier Thermo Scientific Chemicals Supplier No. A1927422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 823-22-3 | C6H10O2 | 114.144 g/mol

d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
d-Hexanolactone is widely used in butter, cream milk, nut, fruit flavors and in cosmetic flavor.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 823-22-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00083574
InChI Key RZTOWFMDBDPERY-UHFFFAOYSA-N
Synonym delta-hexalactone, delta-hexanolactone, delta-caprolactone, 5-methyl-delta-valerolactone, 5-hydroxyhexanoic acid lactone, tetrahydro-6-methyl-2h-pyran-2-one, 2h-pyran-2-one, tetrahydro-6-methyl, .delta.-caprolactone, 6-methylvalerolactone, hexanolactone
PubChem CID 13204
IUPAC Name 6-methyloxan-2-one
SMILES CC1CCCC(=O)O1

Specifications

Melting Point 18°C
Density 1.04
Boiling Point 231°C
Flash Point 105°C (221°F)
Refractive Index 1.451
Quantity 100 g
Beilstein 80501
Solubility Information Immiscible with water.
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material delta-Hexanolactone
Show More Show Less
EINECSNumber 212-511-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only