missing translation for 'onlineSavingsMsg'
Learn More

delta-Dodecanolactone 98.0+%, TCI America™

Catalog No. D141425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D141425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D141425ML Supplier TCI America Supplier No. D141425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 713-95-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00006651
InChI Key QRPLZGZHJABGRS-UHFFFAOYSA-N
Synonym delta-dodecalactone, 5-dodecanolide, delta-dodecanolactone, dodecan-5-olide, 6-heptyltetrahydro-2h-pyran-2-one, 5-dodecalactone, 2h-pyran-2-one, 6-heptyltetrahydro, dodecanolide-1,5, 5-hydroxydodecanoic acid lactone, n-heptyl-delta-valerolactone
PubChem CID 12844
IUPAC Name 6-heptyloxan-2-one
SMILES CCCCCCCC1CCCC(=O)O1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Dodecanolactone
EINECSNumber (5)-0698
RTECSNumber UQ0850000
TSCA Yes