missing translation for 'onlineSavingsMsg'
Learn More

delta-Dodecanolactone, 98%

Catalog No. p-7046459
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1440614 25 g
AAL1440622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1440614 Supplier Thermo Scientific Chemicals Supplier No. L1440614

CAS: 713-95-1 | C12H22O2 | 198.306 g/mol

delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas.

Chemical Identifiers

CAS 713-95-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00006651
InChI Key QRPLZGZHJABGRS-UHFFFAOYSA-N
Synonym delta-dodecalactone, 5-dodecanolide, delta-dodecanolactone, dodecan-5-olide, 6-heptyltetrahydro-2h-pyran-2-one, 5-dodecalactone, 2h-pyran-2-one, 6-heptyltetrahydro, dodecanolide-1,5, 5-hydroxydodecanoic acid lactone, n-heptyl-delta-valerolactone
PubChem CID 12844
IUPAC Name 6-heptyloxan-2-one
SMILES CCCCCCCC1CCCC(=O)O1

Specifications

Melting Point -7°C
Density 0.95
Boiling Point 288°C
Flash Point 165°C (329°F)
Refractive Index 1.46
Quantity 25 g
Beilstein 1282749
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 198.3
Percent Purity 98%
Chemical Name or Material delta-Dodecanolactone
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 211-932-4
RTECSNumber UQ0850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only