Learn More
Microsolv Technology Delivery Cap 3 Hole Top Thred DFS Item
Supplier:  Microsolv Technology Corporation 5882006
Cap, Mobile Phase Delivery System, GL45 Threads, Blue, PE, for HPLC Reservoir Bottles. A PTFE Insert with 3 Holes, PEEK Plug and O-ring is included. MicroSolv Brand. 1/EA.
Catalog No. 5001452650
Only null left
Explore available promotions