Learn More
Thermo Scientific™ Deionizer Cartridges
Catalog No. 1116457
Change view
Click to view available options
For Use With (Equipment):
B-Pure™
Bantam™ Deionizer, B-Pure™
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Resin Type:
Activated Carbon
Extruded Carbon
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Porous Strong Ion Exchange
Strong Cation Exchange
Two Bed Ion Exchange
Ultrapure, Oxygen and Organic Removal
Ultrapure, Oxygen Removal
For Use With (Application):
Cation Removal
High capacity, two-bed ion exchange
Organic Removal
Oxygen Removal
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure, Oxygen and Organic Removal
Capacity:
1760 grains
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
785 grains/1000 gal.
875 grains
30 g
2000 gal.
5000 gal.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Resin Type For Use With (Application) Capacity
1116457 B-Pure™ Mixed Bed Ultrapure -
090343 Bantam™ Deionizer, B-Pure™ Mixed Bed - 875 grains
090348 B-Pure™ Porous Strong Ion Exchange Oxygen Removal 30 g
090345 Bantam™ Deionizer, B-Pure™ Activated Carbon Organic Removal 5000 gal.
0903410 Bantam™ Deionizer, B-Pure™ Strong Cation Exchange Cation Removal 3170 grains
0902833 Bantam™ Deionizer, B-Pure™ Mixed Bed, Activated Carbon Ultrapure and Organic Removal 785 grains/1000 gal.
09034104 B-Pure™ Macroreticular, Activated Carbon Pretreatment 5000 gal.
090345C B-Pure™ Macroreticular, Activated Carbon Pretreatment Macropure 2000 gal.
0905047 B-Pure™ Extruded Carbon Organic Removal 5000 gal.
09050263 B-Pure™ Ultrapure, Oxygen Removal Ultrapure and Organic Removal 455 grains/30 g
0903490 B-Pure™ Ultrapure, Oxygen and Organic Removal Ultrapure, Oxygen and Organic Removal 365 grains/2000 gal./12 g
09034 Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated Two Bed Ion Exchange High capacity, two-bed ion exchange 1760 grains
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 1116457 Supplier Thermo Scientific™ Supplier No. D8816
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
Description High Temp, Ultrapure Cartridge
Resin Type Mixed Bed
For Use With (Application) Ultrapure
For Use With (Equipment) B-Pure™
Type Deionizer Cartridge