missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Deionized Water

Catalog No. STLSL305
Change view
Click to view available options
Quantity:
1 gal./Bottle, 4 Bottles/Cs.
2.5 gal./Bottle
5 gal./Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL305 5 gal./Bottle
STLSL301 1 gal./Bottle, 4 Bottles/Cs.
STLSL3025 2.5 gal./Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STLSL305 Supplier StatLab™ Supplier No. SL305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent ACS grade deionized water, for all standard pathology laboratory applications.

  • Water is purified in-house in the StatLab™ state-of-the-art deionized water purification system
  • Incoming water is filtered and softened before it undergoes a reverse osmosis process
  • Water is further purified by UV, followed by an ultra-purification ion exchange process

Specifications

Color Clear
Grade Reagent
Sterility Non-sterile
Quantity 5 gal./Bottle