missing translation for 'onlineSavingsMsg'
Learn More

Fischer Technical Company™ Dehydrated Culture Media and Ingredients

Catalog No. p-3078969
Change view
Click to view available options
Media Type:
Agar Powder
Tryptic Soy Agar
Quantity:
100 g
454 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Quantity
S70210A Agar Powder 100 g
S71627A Tryptic Soy Agar 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S70210A Supplier Fischer Technical Company™ Supplier No. CD900D
Only null left

Essential agents for preparing culture media.

Successful bacteriological work begins with high-quality media.

  • Agar, a solidifying agent, is an essential component of many media
  • Available in a variety of formulations and bottle sizes
  • For use in preparing plated media, tubed media, bottled media, and slants

Specifications

Form Dehydrated
Format Bottled
Manufacturing Origin US with Globally Sourced Materials
Media Type Agar Powder
Product Type Microbiology Media
Sterility Non-sterile
Target Organism Class Bacteria
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 100 g