Learn More
Fisherbrand™ DEHP-Free PVC Tubing
GREENER_CHOICE
Manufactured from the highest-grade resin materials and free from ortho-phthalate plasticizers. Resists hardening and discoloration from a wide range of laboratory chemicals. Ideal for a wide range of non-medical, general lab, research, and pharmaceutical applications.
Supplier:  Fisherbrand™ 14387345
 View more versions of this product
Catalog No. 14387345
Only null left
Description
Description
Fisherbrand™ DEHP-Free Clear Plastic PVC Tubing is manufactured from the highest-grade resin materials and free from ortho-phthalate plasticizers. Resists hardening and discoloration from a wide range of laboratory chemicals and does not impart odor or taste making this tubing ideal for a wide range of non-medical, general lab, research and pharmaceutical applications.
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Free from ortho-phthalate plasticizers
  • Resists hardening and discoloration from a wide range of chemicals - demonstrates dimensional stability
  • Marked with I.D. and regulatory compliance as well as at 12 in. intervals for easy identification and measurement
  • Durometer hardness is Shore A, 60
  • Does not impart odor or taste
  • Can write on with Lab marker
  • Slips easily over glass and plastic fittings; grips securely
  • Autoclavable/Transparent
Specifications
Chemical Identifiers
Autoclavable
50 ft.
23°C
Non-sterile
Coil
Non-Medical
60 (Shore A)
160°F
PVC Tubing
PVC
Can be used for Fluid and Gas Phase Material Transfer in General Lab, Research and Pharmaceutical Facilities
Acids - Weak; Bases - Weak; Alcohol - Aliphatic; Bases - Strong
Autoclave, Gas, Chemical
Room temperature, avoid direct sunlight
Clear
Flexible
71°C
Specifications
Autoclavable
PVC
50 ft.
Can be used for Fluid and Gas Phase Material Transfer in General Lab, Research and Pharmaceutical Facilities
Acids - Weak; Bases - Weak; Alcohol - Aliphatic; Bases - Strong
Autoclave, Gas, Chemical
Room temperature, avoid direct sunlight
Clear
Flexible
71°C
1/16 in.
1/8 in.
1/32 in.
23°C
Non-sterile
Coil
Non-Medical
60 (Shore A)
160°F
PVC Tubing
Product Suggestions
Product Suggestions