missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deflazacort 98.0+%, TCI America™
$391.44 - $1168.76
Chemical Identifiers
| CAS | 14484-47-0 |
|---|---|
| Molecular Formula | C25H31NO6 |
| Molecular Weight (g/mol) | 441.524 |
| MDL Number | MFCD00866106 |
| InChI Key | FBHSPRKOSMHSIF-GRMWVWQJSA-N |
| PubChem CID | 189821 |
| SMILES | CC1=NC2(C(O1)CC3C2(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)C(=O)COC(=O)C |
Chemical Identifiers
| 14484-47-0 | |
| 441.524 | |
| FBHSPRKOSMHSIF-GRMWVWQJSA-N | |
| CC1=NC2(C(O1)CC3C2(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)C(=O)COC(=O)C |
| C25H31NO6 | |
| MFCD00866106 | |
| 189821 |
Specifications
| 14484-47-0 | |
| White | |
| MFCD00866106 | |
| FBHSPRKOSMHSIF-GRMWVWQJSA-N | |
| 441.524 | |
| 441.52 | |
| Crystalline Powder |
| 258°C | |
| C25H31NO6 | |
| 1 g | |
| CC1=NC2(C(O1)CC3C2(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)C(=O)COC(=O)C | |
| 189821 | |
| ≥98.0% (HPLC,N) | |
| Deflazacort |
Safety and Handling
RTECSNumber : TU4157050
TSCA : No
Recommended Storage : Refrigerator