missing translation for 'onlineSavingsMsg'
Learn More

Deflazacort 98.0+%, TCI America™

Catalog No. D45231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45231G 1 g
D45235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45231G Supplier TCI America Supplier No. D45231G
Only null left

Chemical Identifiers

CAS 14484-47-0
Molecular Formula C25H31NO6
Molecular Weight (g/mol) 441.524
MDL Number MFCD00866106
InChI Key FBHSPRKOSMHSIF-GRMWVWQJSA-N
PubChem CID 189821
SMILES CC1=NC2(C(O1)CC3C2(CC(C4C3CCC5=CC(=O)C=CC45C)O)C)C(=O)COC(=O)C

Specifications

Melting Point 258°C
Color White
Quantity 1 g
Formula Weight 441.52
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Deflazacort
RTECSNumber TU4157050
TSCA No
Recommended Storage Refrigerator