missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Defibrinated Sheep Blood

Catalog No. R54008
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R54008 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R54008 Supplier Thermo Scientific™ Supplier No. NTB0016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Be confident that your animal blood meets the highest standards possible by using Defibrinated Sheep Blood, sourced from our own technologically-advanced sheep farm.

Be confident that your animal blood meets the highest standards possible by using Thermo Scientific™ Defibrinated Sheep Blood, sourced from our own technologically-advanced sheep farm.

Specifications

Species Sheep
Form Liquid
Product Type Whole Blood
Quantity 50 mL
Processing Defibrinated