missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Deepwell Plates BioBased

Catalog No. 14278170
Change view
Click to view available options
Color:
Green
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14278170 White
14278175 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14278170 Supplier Eppendorf™ Supplier No. 0030501012

Deepwell Plates BioBased support all manual and automated workflows, from -86°C sample storage to +100°C DNA denaturation. Featuring OptiTrack labeling and a stable, durable design, they ensure reliable robotic handling and simplify identification in high-throughput labs.

  • Two different colored rims available for easy process differentiation.
  • Consistent geometry ensures smooth integration with robotic applications.
  • Laser-engraved lot number and expiry date on each plate for traceability.
  • Raised well rims and smooth surfaces provide reliable heat sealing and simple stacking.
  • RecoverMax™ well design enhances sample recovery and ensures efficient mixing.
  • Minimized residual volume and high well-to-well uniformity deliver reproducible results.
  • Premium polypropylene construction resists chemicals, mechanical stress, and extreme conditions.
  • Free from slip agents, plasticizers, and biocides that may interfere with bioassays.
  • Autoclavable at 121°C for 20 minutes, ensuring sterility and reuse when needed.
  • High-contrast OptiTrack™ matrix improves sample identification speed and reduces pipetting errors.

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 20 plates (5 bags x 4 plates)
Color White
Product Type Deepwell Plate
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Volume (Metric) Working 50 to 2000 μL
Sterility Non-sterile
Surface Treatment Non-coated
Well Shape Round
Dnase Free Yes
For Use With (Application) Sample storage and preparation, bacteria and yeast cultivation, DNA and RNA isolation with high throughput
Green Features Biobased, ACT Label
Max. RCF 6000 x g
No. per Pack 4
Purity or Quality Grade PCR clean and Protein-free
Quantity 20
RNase Free Yes
Well Color Clear
Dimensions (L x W x H) 85.5 x 127.8 x 60 mm
Fit for Purpose PCR Ready
Show More Show Less