missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethylammonium Chloride 98.0+%, TCI America™

Catalog No. D130625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D130625G Supplier TCI America Supplier No. D130625G

Chemical Identifiers

CAS 10108-87-9
Molecular Formula C13H30ClN
Molecular Weight (g/mol) 235.84
MDL Number MFCD00059967
InChI Key HXWGXXDEYMNGCT-UHFFFAOYSA-M
PubChem CID 24951
IUPAC Name decyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Color White
Quantity 25 g
Formula Weight 235.84
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Decyltrimethylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
TSCA No