missing translation for 'onlineSavingsMsg'
Learn More

Decyltrimethoxysilane 97.0+%, TCI America™

Catalog No. p-7128415
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D470425G 25 g
D4704250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D470425G Supplier TCI America Supplier No. D470425G
Only null left

Chemical Identifiers

CAS 5575-48-4
Molecular Formula C13H30O3Si
Molecular Weight (g/mol) 262.47
MDL Number MFCD00069135
InChI Key KQAHMVLQCSALSX-UHFFFAOYSA-N
Synonym n-decyltrimethoxysilane, decyltrimethoxysilan, trimethoxydecylsilane, decyl trimethoxy silane, silane,decyltrimethoxy, decyl-trimethoxysilane
PubChem CID 2758023
IUPAC Name decyltrimethoxysilane
SMILES CCCCCCCCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 g
Formula Weight 262.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyltrimethoxysilane
EINECSNumber (2)-3512
TSCA No