missing translation for 'onlineSavingsMsg'
Learn More

Decyltrichlorosilane 98.0+%, TCI America™

Catalog No. D148625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D148625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D148625ML Supplier TCI America Supplier No. D148625ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13829-21-5
Molecular Formula C10H21Cl3Si
Molecular Weight (g/mol) 275.713
MDL Number MFCD00013600
InChI Key HLWCOIUDOLYBGD-UHFFFAOYSA-N
Synonym decyltrichlorosilane, trichloro decyl silane, n-decyltrichlorosilane, silane, trichlorodecyl, acmc-209chi, trichloro n-decyl silane, trichloro decyl silane #
PubChem CID 83764
IUPAC Name trichloro(decyl)silane
SMILES CCCCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 mL
UN Number 2987
Formula Weight 275.71
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyltrichlorosilane
EINECSNumber (2)-2041
TSCA Yes