missing translation for 'onlineSavingsMsg'
Learn More

Decyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. p-7128178
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42465G 5 g
D424625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42465G Supplier TCI America Supplier No. D42465G
Only null left

Chemical Identifiers

CAS 15163-36-7
Molecular Formula C15H33NO3S
Molecular Weight (g/mol) 307.49
MDL Number MFCD00036908
InChI Key WKALLSVICJPZTM-UHFFFAOYSA-N
Synonym 3-decyldimethylammonio propane-1-sulfonate, n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, sulfobetaine 10, caprylyl sulfobetaine, 3-decyldimethylammonio propanesulfonate, zwittergent 3-10 detergent, capryl sultaine, zwittergent 310, 3-decyl dimethyl ammonio propane-1-sulfonate, 3-decyldimethylamino propanesulfonic acid
PubChem CID 161111
IUPAC Name 3-(decyldimethylazaniumyl)propane-1-sulfonate
SMILES CCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Melting Point 229°C
Color White
Quantity 5 g
Formula Weight 307.49
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Decyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]