missing translation for 'onlineSavingsMsg'
Learn More

Decylbenzene 98.0+%, TCI America™

Catalog No. D00345ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00345ML 5 mL
D003425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00345ML Supplier TCI America Supplier No. D00345ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-72-3
Molecular Formula C16H26
Molecular Weight (g/mol) 218.38
MDL Number MFCD00008955
InChI Key UZILCZKGXMQEQR-UHFFFAOYSA-N
Synonym 1-phenyldecane, benzene, decyl, n-decylbenzene, decane, 1-phenyl, n-decyl benzene, unii-kir29w0bp0, kir29w0bp0, phenyldecane, hsdb 920, decane, phenyl
PubChem CID 7716
IUPAC Name decylbenzene
SMILES CCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -14°C
Color Colorless
Boiling Point 298°C
Quantity 5 mL
Formula Weight 218.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decylbenzene
EINECSNumber (3)-0021
TSCA Yes