missing translation for 'onlineSavingsMsg'
Learn More

Decyl Decanoate 95.0+%, TCI America™

Catalog No. D108025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D108025ML 25 mL
D1080500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D108025ML Supplier TCI America Supplier No. D108025ML
Only null left

Chemical Identifiers

CAS 1654-86-0
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00059280
InChI Key XAKXZZPEUKNHMA-UHFFFAOYSA-N
Synonym decanoic acid, decyl ester, decanoic acid decyl ester, unii-b88b7acl5l, b88b7acl5l, n-capric acid n-decyl ester, capryl caprate, acmc-209dsk, decanoic acid, decylester
PubChem CID 74247
IUPAC Name decyl decanoate
SMILES CCCCCCCCCCOC(=O)CCCCCCCCC

Specifications

Color Yellow
Boiling Point 159°C
Quantity 25 mL
Formula Weight 312.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Decyl Decanoate
EINECSNumber (2)-0782&(2)-2488
TSCA Yes