missing translation for 'onlineSavingsMsg'
Learn More

Decyl decanoate, 98%, Thermo Scientific™

Catalog No. AC457110250
Change view
Click to view available options
Quantity:
25mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457110250 25mL
AC457115000 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457110250 Supplier Thermo Scientific Chemicals Supplier No. 457110250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1654-86-0
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
InChI Key XAKXZZPEUKNHMA-UHFFFAOYSA-N
Synonym decanoic acid, decyl ester, decanoic acid decyl ester, unii-b88b7acl5l, b88b7acl5l, n-capric acid n-decyl ester, capryl caprate, acmc-209dsk, decanoic acid, decylester
PubChem CID 74247
IUPAC Name decyl decanoate
SMILES CCCCCCCCCCOC(=O)CCCCCCCCC

Specifications

Density 0.8590g/mL
Boiling Point 174.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)8CO2(CH2)9CH3
Quantity 25mL
Specific Gravity 0.859
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 312.54
Percent Purity 98%
Chemical Name or Material Decyl decanoate
Show More Show Less
EINECSNumber 216-729-4

RUO – Research Use Only