missing translation for 'onlineSavingsMsg'
Learn More

Decyl Chloroformate 95.0+%, TCI America™

Catalog No. c135725ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
Catalog No. Quantity
C135725ML 25 mL
1 options

Catalog No. C135725ML

Supplier: TCI America C135725ML

Only null left

Chemical Identifiers

CAS 55488-51-2
Molecular Formula C11H21ClO2
Molecular Weight (g/mol) 220.737
MDL Number MFCD00126853
InChI Key AZZCHVHSWUYCQA-UHFFFAOYSA-N
Synonym Chloroformic Acid Decyl Ester
PubChem CID 108727
IUPAC Name decyl carbonochloridate
SMILES CCCCCCCCCCOC(=O)Cl

Specifications

Color Colorless
Boiling Point 163°C
Quantity 25 mL
UN Number 3277
Formula Weight 220.74
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Decyl Chloroformate