missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Proguard™ Professional Hand Cream
GREENER_CHOICE

Catalog No. 0435521
Change view
Click to view available options
Volume (Metric):
90 mL
473.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
0435521 473.1 mL
0435520 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0435521 Supplier Decon™ Supplier No. 9516
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation

  • Does not contain grease, oil, perfume, aloe, or other ingredients that can compromise the integrity of latex gloves
  • Absorbs instantly, leaving skin soft

Specifications

Product Type Professional Hand Cream
Color White
Form Cream
For Use With (Application) Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation
Latex Free Yes
Packaging Pump
Quantity 1 Ea.
Volume (English) 16 oz.
Volume (Metric) 473.1 mL