missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Jet Stream™ High-Pressure Dust Remover
GREENER_CHOICE

Catalog No. 22281495
Change view
Click to view available options
Quantity:
11 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22281495 11 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22281495 Supplier Decon™ Supplier No. 3711
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Removes microscopic dust from laboratory equipment

Ideal for use on negatives, microscope slides, lenses and balances.

  • Moisture-free

Specifications

Product Type High Pressure Dust Remover
Color-coded No
Form Aerosol
Scent Unscented
Sterility Non-sterile
Flammable No
For Use With (Application) Ideal for use on negatives, microscope slides, lenses and balances
Quantity 11 oz.
Volume (English) 11 oz.
Volume (Metric) 312 g