missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Gooseneck Trigger Sprayers

Catalog No. 0435612
Change view
Click to view available options
Mounting:
Freestanding
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
0435612 Freestanding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0435612 Supplier Decon™ Supplier No. 8309NS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Spray solutions directly from one gallon containers

  • Attached to flexible tubing that extends 22 in. from container
  • For use with Decon's 1G CiDehol 70 product – Decon #8401

Specifications

Product Type Manual Dispenser
Color Clear, White
Length (Metric) 20.3 cm
Holds Hand Soap
Material Plastic
Mounting Freestanding
No. of Compartments 1
Product Style Open Top
Sterility Non-sterile