missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decon™ Dri-Contrad™ Detergent Powder

Cleans proteins, bloods, oils, waxes, and organic or inorganic contaminants
Supplier: Decon™ DC2007
Description
- Nonfoaming powder
- For automatic washers
- Free-rinsing, rapidly soluble, and noncorrosive
- Chlorinated for biohazard control
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Chemical Identifiers
| Detergent | |
| Powder | |
| Non-sterile |
| No | |
| Regular Scent | |
| For automatic washers, removes Proteins, Blood, Oils, Waxes and Organic or Inorganic contaminants |
Specifications
| Detergent | |
| Powder | |
| Non-sterile | |
| For automatic washers, removes Proteins, Blood, Oils, Waxes and Organic or Inorganic contaminants | |
| 27.5 lb. |
| No | |
| Regular Scent | |
| Poly Bottle | |
| 12.5 kg |